Learn More
Invitrogen™ TUFM Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA592964
Description
Immunogen sequence: ADAVQDSEMV ELVELEIREL LTEFGYKGEE TPVIVGSALC ALEGRDPELG LKSVQKLLDA VDTYIPVPAR DLEKPFLLPV EAVYSVPGRG TVVTGTLERG ILKKGDECEL LGHSKNIRTV VTGIEMFHKS LERAEAGDNL GALVRGLKRE DLRRGLVMVK PGSIKPHQKV EAQVYILSKE EGGRHKPFVS HFMPVMFSLT WDMACRIILP PEKELAMPGE DLKFNLILRQ PMILEKGQRF TLRDGNRTIG TGLVTNTLAM TEEEKNIKWG; Positive Samples: HT-29, HepG2, HeLa, A-431, Jurkat, Mouse kidney, Mouse liver, Rat kidney, Rat liver; Cellular Location: Mitochondrion.
This gene encodes a protein which participates in protein translation in mitochondria. Mutations in this gene have been associated with combined oxidative phosphorylation deficiency resulting in lactic acidosis and fatal encephalopathy. A pseudogene has been identified on chromosome 17.
Specifications
TUFM | |
Polyclonal | |
Unconjugated | |
TUFM | |
2300002G02Rik; C76308; C76389; COXPD4; D250; EFTU; EF-Tu; EF-TuMT; elongation factor Tu; elongation factor Tu, mitochondrial; fi06f04; P43; Tu translation elongation factor, mitochondrial; TUFM; tufm {ECO:0000250; UniProtKB:P49411}; wu:fi06f04; zgc:110766 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
233870, 293481, 7284 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot, Immunocytochemistry | |
1.31 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P49411, P85834, Q8BFR5 | |
TUFM | |
Recombinant fusion protein containing a sequence corresponding to amino acids 186-455 of human TUFM (NP_0033123). | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.