Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ TUG 891
GREENER_CHOICESDP

Catalog No. 460150 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
46-015-0 50 mg
46-011-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 46-015-0 Supplier Tocris Bioscience™ Supplier No. 4601/50
Add to Cart
Add to Cart
This item is not returnable. View return policy

Potent and selective FFA4 (GPR120) agonist

Potent FFA4 (GPR120) agonist (pEC50 values are 7.36 and 7.77 for human and mouse GPR120 respectively). Selective for GPR120 over free fatty acid receptors (pEC50 = 4.19 for FFA1; displays no activity at FFA2 or FFA3). Cell permeable.

Chemical Identifiers

CAS 1374516-07-0
Molecular Formula C23H21FO3

Specifications

Quantity 50 mg
Formula Weight 364.41
Purity >99%
Molecular Weight (g/mol) 364.416
Chemical Name or Material TUG 891
Recommended Storage Store at Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.