Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TUG Antibody (SR2139), Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP322469100UL
Description
TUG Monoclonal antibody specifically detects TUG in Human, Mouse, Rat samples. It is validated for Western Blot, ImmunohistochemistrySpecifications
TUG | |
Monoclonal | |
Unconjugated | |
PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human, Mouse, Rat | |
Purified |
Western Blot, Immunohistochemistry | |
SR2139 | |
Western Blot 1:500-1:2000, Immunohistochemistry 1:50-1:200 | |
Alveolar soft part sarcoma chromosomal region candidate gene 1 protein, alveolar soft part sarcoma chromosome region, candidate 1, Alveolar soft part sarcoma locus, ASPLUBX domain-containing protein 9, ASPSrenal cell carcinoma gene on chromosome 17, RCC17renal cell carcinoma, papillary, 17, Renal papillary cell carcinoma protein 17, tether containing UBX domain for GLUT4, TUG, UBX domain protein 9, UBXD9ASPCR1, UBXN9FLJ45380 | |
A synthesized peptide derived from human TUG (Uniprot #: Q9BZE9) | |
100 μg | |
Cancer, Cell Biology, Endocrinology, Signal Transduction | |
79058 | |
Store at -20° C. Avoid freeze/thaw cycles. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction