Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Tug Monoclonal Antibody (4A11A6G11)
GREENER_CHOICE

Catalog No. PIMA515269
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA515269 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA515269 Supplier Invitrogen™ Supplier No. MA515269
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MA5-15269 targets Mouse TUG in indirect ELISA, WB applications and shows reactivity with mouse samples. The MA5-15269 immunogen is purified recombinant fragment of TUG expressed in E. Coli.

This gene is a candidate gene for alveolar soft part sarcoma (ASPS). It has been found that this gene is fused with transcription factor TFE3 gene in ASPS and also in renal cell carcinomas. Several alternatively spliced transcript variants of this gene have been described, but their full length nature has not been determined.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Tug
Applications ELISA, Western Blot
Classification Monoclonal
Clone 4A11A6G11
Concentration Conc. Not Determined
Conjugate Unconjugated
Formulation Ascites with 0.03% sodium azide
Gene ASPSCR1
Gene Accession No. Q8VBT9
Gene Alias 1190006K01Rik; alveolar soft part sarcoma chromosomal region candidate gene 1 protein; Alveolar soft part sarcoma chromosomal region candidate gene 1 protein homolog; alveolar soft part sarcoma chromosome region, candidate 1; alveolar soft part sarcoma chromosome region, candidate 1 (human); alveolar soft part sarcoma locus; ASPC; ASPCR1; ASPL; ASPS; Aspscr1; ASPSCR1, UBX domain containing tether for SLC2A4; RCC17; renal cell carcinoma gene on chromosome 17; renal cell carcinoma, papillary, 17; renal papillary cell carcinoma protein 17; Tether containing UBX domain for GLUT4; tether, containing a UBX domain, for GLUT4; TUG; UBX domain protein 9; UBX domain-containing protein 9; UBXD9; UBXN9
Gene Symbols ASPSCR1
Host Species Mouse
Immunogen Purified recombinant fragment of TUG expressed in E. Coli.
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 68938
Target Species Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.