Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Tug Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA596695
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA596695 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA596695 Supplier Invitrogen™ Supplier No. PA596695
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: KSKSGQDPQQ EQEQERERDP QQEQERERPV DREPVDREPV VCHPDLEERL QAWPAELPDE FFELTVDDVR RRLAQLKSER KRLEEAPLVT KAFREAQIKE KLERYPKVAL RVLFPDRYVL QGFFRPSETV GDLRDFVRSH LGNPELSFYL FITPPKTVLD DHTQTLFQAN LFPAALVHLG AEEPAGVYLE PGLLEHAISP SAADVLVARY MSRAAGSPSP LPAPDPAPKS EPAAEEGALV PPEPIPGTAQ PVKRSLGKVP KWLKLPASKR; Positive Samples: 293T, SW480, A-549, BT-474, Mouse liver, Mouse testis; Cellular Location: Cytoplasm, Endomembrane system, Endoplasmic reticulum-Golgi intermediate compartment membrane, Nucleus, Peripheral membrane protein.

This gene is a candidate gene for alveolar soft part sarcoma (ASPS). It has been found that this gene is fused with transcription factor TFE3 gene in ASPS and also in renal cell carcinomas. Several alternatively spliced transcript variants of this gene have been described, but their full length nature has not been determined.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Tug
Applications ELISA, Immunoprecipitation, Western Blot, Immunocytochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 8.71 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene ASPSCR1
Gene Accession No. Q8VBT9, Q9BZE9
Gene Alias 1190006K01Rik; alveolar soft part sarcoma chromosomal region candidate gene 1 protein; Alveolar soft part sarcoma chromosomal region candidate gene 1 protein homolog; alveolar soft part sarcoma chromosome region, candidate 1; alveolar soft part sarcoma chromosome region, candidate 1 (human); alveolar soft part sarcoma locus; ASPC; ASPCR1; ASPL; ASPS; Aspscr1; ASPSCR1, UBX domain containing tether for SLC2A4; RCC17; renal cell carcinoma gene on chromosome 17; renal cell carcinoma, papillary, 17; renal papillary cell carcinoma protein 17; Tether containing UBX domain for GLUT4; tether, containing a UBX domain, for GLUT4; TUG; UBX domain protein 9; UBX domain-containing protein 9; UBXD9; UBXN9
Gene Symbols ASPSCR1
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 284-553 of human TUG/ASPSCR1 (NP_0769881).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 68938, 691026, 79058
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.