Learn More
Invitrogen™ Tug Recombinant Rabbit Monoclonal Antibody (8I0U8)

Description
Positive test controls include: 293T, NIH/3T3, RD, Mouse testis. The target is usually found in the following locations: Cytoplasm, Endomembrane system, Endoplasmic reticulum-Golgi intermediate compartment membrane, Nucleus, Peripheral membrane protein. Immunogen sequence: LPLESGELSR GDLSRPEDAD TSGPCCEHTQ EKQSTRAPAA APFVPFSGGG QRLGGPPGPT RPLTSSSAKL PKSLSSPGGP SKPKKSKSGQ DPQQEQEQER E.
Specifications
Specifications
| Antigen | Tug |
| Applications | ELISA, Western Blot |
| Classification | Recombinant Monoclonal |
| Clone | 8I0U8 |
| Concentration | 0.24 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.05% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 |
| Gene | ASPSCR1 |
| Gene Accession No. | Q8VBT9, Q9BZE9 |
| Gene Alias | 1190006K01Rik; alveolar soft part sarcoma chromosomal region candidate gene 1 protein; Alveolar soft part sarcoma chromosomal region candidate gene 1 protein homolog; alveolar soft part sarcoma chromosome region, candidate 1; alveolar soft part sarcoma chromosome region, candidate 1 (human); alveolar soft part sarcoma locus; ASPC; ASPCR1; ASPL; ASPS; Aspscr1; ASPSCR1, UBX domain containing tether for SLC2A4; RCC17; renal cell carcinoma gene on chromosome 17; renal cell carcinoma, papillary, 17; renal papillary cell carcinoma protein 17; Tether containing UBX domain for GLUT4; tether, containing a UBX domain, for GLUT4; TUG; UBX domain protein 9; UBX domain-containing protein 9; UBXD9; UBXN9 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.