Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TULA/STS-2 Antibody, PerCP, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP300200PCP
Description
TULA/STS-2 Polyclonal antibody specifically detects TULA/STS-2 in Human samples. It is validated for ELISA, Immunohistochemistry (Paraffin)Specifications
TULA/STS-2 | |
Polyclonal | |
ELISA, Immunohistochemistry-Paraffin | |
Cbl-interacting protein 4, gene similar to UBA containing SH3 domain10CLIP4STS-2T-cell ubiquitin ligand protein, STS2, Suppressor of T-cell receptor signaling 2, T-cell ubiquitin ligand, TULAubiquitin-associated and SH3 domain-containing protein A, ubiquitin associated and SH3 domain containing A | |
Produced in rabbits immunized with purified, recombinant Human TULA/STS-2 (Accession#: AAH69511.1; Ala354-Asn623) | |
0.1 mL | |
Primary | |
Human | |
Purified |
ELISA, Immunohistochemistry (Paraffin) | |
PerCP | |
PBS | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
53347 | |
Store at 4C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction