Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Tulobuterol

Catalog No. AAJ6397503
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ6397503 1 g
AAJ6397506 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ6397503 Supplier Thermo Scientific™ Supplier No. J6397503

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

beta-2-adrenergic receptor agonist

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

Quantity 1 g
Description White to Off-white
Content And Storage -30°C to -10°C
For Use With (Application) beta-2-adrenergic receptor agonist
CAS 41570-61-0
Molecular Formula C12H18ClNO
Formula Weight 227.73g/mol
Melting Point 159°C to 163°C
Product Type Signal Transduction Reagent
Form Crystalline Powder
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.