Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TULP2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA541449
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA541449 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA541449 Supplier Invitrogen™ Supplier No. PA541449
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: MSQDNDTLMR DILGHELAAM RLQKLEQQRR LFEKKQRQKR QELLMVQANP Sequence homology: Cow: 86%; Dog: 79%; Guinea Pig: 79%; Horse: 93%; Human: 100%; Mouse: 86%; Rat: 86%.

TULP2 is a member of a family of tubby-like genes (TULPs) that encode proteins of unknown function. Members of this family have been identified in plants, vertebrates, and invertebrates. The TULP proteins share a conserved C-terminal region of approximately 200 amino acid residues.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TULP2
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene TULP2
Gene Accession No. O00295
Gene Alias cancer testis antigen 65; cancer/testis antigen 65; CT65; Pdet; Protein P4-6; TUB like protein 2; tubby like protein 2; tubby-like protein 2; Tubby-related protein 2; TUBL2; TULP2
Gene Symbols TULP2
Host Species Rabbit
Immunogen Synthetic peptide directed towards the N-terminal of human TULP2 (aa 1-50).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7288
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.