Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TULP3 Antibody (OTI7B8), Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP201671
Description
TULP3 Monoclonal specifically detects TULP3 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
TULP3 | |
Monoclonal | |
0.83 mg/mL | |
Western Blot 1:2000, Flow Cytometry 1:100, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:100, Immunohistochemistry-Paraffin 1:150 | |
TULP3 | |
Full length human recombinant protein of human TULP3(NP_003315) produced in HEK293T cell. | |
Affinity Purified | |
RUO | |
Primary | |
Human, Monkey | |
IgG1 |
Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
OTI7B8 | |
Unconjugated | |
MGC45295, tubby like protein 3, tubby-related protein 3, TUBL3Tubby-like protein 3 | |
Mouse | |
49.5 kDa | |
0.1 mL | |
Signal Transduction | |
7289 | |
Store at -20°C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction