Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TULP3 Mouse anti-Human, Clone: OTI4E5, liquid, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene TA504149

Description
This gene encodes a member of the tubby-like protein (TULP) family. Members of this family have been identified in plants, vertebrates, and invertebrates, and they share a conserved C-terminal region of approximately 200 amino acid residues. The human and mouse TULP3 proteins share 69% amino acid sequence identity, with higher identity at the N and C termini than in the central region.Specifications
TULP3 | |
Monoclonal | |
0.78 mg/mL | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
O75386 | |
TULP3 | |
Full length human recombit protein of human TULP3 produced in HEK293T cell. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG1 |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
OTI4E5 | |
Unconjugated | |
TULP3 | |
TUBL3 | |
Mouse | |
Affinity Chromatography | |
RUO | |
7289 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction