Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TULP3 Antibody (OTI7B8), FITC, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP274721F
Description
TULP3 Monoclonal specifically detects TULP3 in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.Specifications
TULP3 | |
Monoclonal | |
FITC | |
PBS with 0.05% Sodium Azide | |
TULP3 | |
Full length recombinant protein of human TULP3 (NP_003315) produced in HEK293T cells. | |
0.1 mL | |
Primary | |
Human | |
IgG1 |
Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
OTI7B8 | |
Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
MGC45295, tubby like protein 3, tubby-related protein 3, TUBL3Tubby-like protein 3 | |
Mouse | |
Affinity Purified | |
Signal Transduction | |
7289.0 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction