Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TULP3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA564973
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA564973 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA564973 Supplier Invitrogen™ Supplier No. PA564973
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: QAKLDYQRLLL EKRQRKKRLE PFMVQPNPEA RLRRAKPRAS DEQTPLVNCH TPHSNVILHG IDGPAAVLKP DEVHAPSVSS SVVEEDAENT VDTASKPGLQ ERLQKHDISE SVNFDEETDG ISQSACLERP NSASSQ Highest antigen sequence identity to the following orthologs - mouse 54%, rat 56%.

This gene encodes a member of the tubby-like protein (TULP) family. Members of this family have been identified in plants, vertebrates, and invertebrates, and they share a conserved C-terminal region of approximately 200 amino acid residues. The human and mouse TULP3 proteins share 69% amino acid sequence identity, with higher identity at the N and C termini than in the central region.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TULP3
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene TULP3
Gene Accession No. O75386
Gene Alias 2310022L06Rik; AI316887; TUB like protein 3; tubby like protein 3; tubby-like protein 3; Tubby-related protein 3; TUBL3; Tulp3
Gene Symbols TULP3
Host Species Rabbit
Immunogen Recombinant Human TULP3. Recombinant protein control fragment (Product #RP-105745).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7289
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.