Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tumor-Associated Glycoprotein (TAG-72) Monoclonal Antibody (B72.3), Abbomax

Catalog No. 502204194
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-220-4194 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-220-4194 Supplier Abbomax Inc Supplier No. 603880
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Positive control: Breast Carcinoma Cellular location: Cytoplasmic.

Tumor-Associated Glycoprotein (TAG-72) Monoclonal antibody specifically detects Tumor-Associated Glycoprotein (TAG-72) in Human samples. It is validated for Immunohistochemistry (Paraffin), Western Blot

Specifications

Antigen Tumor-Associated Glycoprotein (TAG-72)
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone B72.3
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with proprietary stabilizer and 0.01% sodium azide
Gene Alias TAG-72; Tumor-associated glycoprotein 72
Host Species Mouse
Immunogen A membrane extracts of human hepatic carcinoma
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.