Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tungsten boride, 99.4% (metals basis)

Catalog No. p-7051624
Click to view available options
Quantity:
25 g
100 g

CAS: 12007-98-6 | B5W2 | 421.73 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12007-98-6
Molecular Formula B5W2
Molecular Weight (g/mol) 421.73
MDL Number MFCD00049707
InChI Key ZFRNSGNJMJAUMG-UHFFFAOYSA-N
Synonym ditungsten pentaboride, w2b5, diboranylidyne-1,2,4-tribora-3,5-ditungstacyclopenta-1,3-diene
PubChem CID 25113386
IUPAC Name boranylidyne($l^{2}-boranylidene)tungsten;boranylidynetungsten;$l^{2}-boranylideneboron
SMILES [B]=[B].B#[W].B#[W]=[B]

Specifications

Physical Form Powder
Quantity 25 g
Solubility Information Insoluble in water.
Formula Weight 421.76
Percent Purity 99.4%
Odor Odorless
Assay (metals basis)
Chemical Name or Material Tungsten boride
EINECSNumber 234-524-8
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.