Alfa Aesar™ Tungsten boride, 99% (metals basis)

Manufacturer: Alfa Aesar™ 1255914

Quantity 25g

 View more versions of this product

Catalog No. AA1255914

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 12007-09-9
    Formula Weight 194.66
    MDL Number MFCD00049708
    Chemical Name or Material Tungsten boride

    Specifications

    Formula Weight 194.66
    TSCA TSCA
    Odor Odorless
    Physical Form Powder
    CAS 12007-09-9
    MDL Number MFCD00049708
    Quantity 25g
    Chemical Name or Material Tungsten boride
    Assay 99%, (metals basis)

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit