Tungsten dichloride dioxide, 99%, Alfa Aesar™

Manufacturer: Alfa Aesar 8996218

Quantity 50g

 View more versions of this product

Catalog No. AA8996218

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 13520-76-8
    Physical Form Powder
    Assay Percent Range 99%
    Molecular Formula WO2Cl2
    Formula Weight 286.75
    MDL Number MFCD00054136

    Specifications

    Formula Weight 286.75
    Quantity 50g
    Color Light yellow
    Physical Form Powder
    CAS 13520-76-8
    Odor Acrid
    Assay Percent Range 99%
    Molecular Formula WO2Cl2
    MDL Number MFCD00054136

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit