Learn More
Tungsten foil, 0.05mm (0.002in) thick, 99.95% (metals basis)
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Quantity | 100 x 100 mm |
Weight | ≈2.4g/50x50mm |
Form | Foil |
Percent Purity | 99.95% |
Assay | (metals basis) |
Chemical Name or Material | Tungsten foil, 0.05mm (0.002 in.) thick |
Frequently Asked Questions (FAQs)
The Tungsten foil, 0.05mm (0.002in) thick, 99.95% (metals basis) may have an oil or oxide coating. To remove the coating, we recommend soaking the foil in very hot water with strong detergent for 10-15 minutes. Wipe the foil with a sponge. Then rinse it with water and dry. This process will remove the coating.
Safety and Handling
EINECSNumber | 231-143-9 |
TSCA | Yes |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.