Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tungsten ICP Standard, 10,000 ppm W in 5% HNO3, Ricca Chemical
SDP

Catalog No. PW10KN100 Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
50 mL
100 mL
Packaging:
Natural LDPE Bottle
Natural Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
PW10KN-100 100 mL Natural LDPE Bottle
PW10KN-50 50 mL Natural Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. PW10KN-100 Supplier Ricca Chemical Company Supplier No. PW10KN100
Add to Cart
Add to Cart

Chemical Identifiers

CAS 7440-33-7 , 7664-39-3 , 7664-39-3 , 7440-33-7
Molecular Formula W
Molecular Weight (g/mol) 183.84
MDL Number MFCD00011461
InChI Key WFKWXMTUELFFGS-UHFFFAOYSA-N
PubChem CID 23964
ChEBI CHEBI:27998
IUPAC Name tungsten
SMILES [W]

Specifications

Packaging Natural LDPE Bottle
Concentration or Composition Notes 10,000ppm W
CAS Min % 82.0, 11.09, 4.1, 0.82
CAS Max % 85.0, 11.54, 4.27, 0.85
Equivalence 1 mL = 10 mg W
Physical Form Liquid
Color Colorless
pH < 2
Type Standard
Quantity 100 mL
Chemical Name or Material Tungsten ICP Standard
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.