Learn More
Tungsten(IV) oxide, 99.9% (metals basis)
Description
Tungsten(IV) oxide is used in the production of tungsten and tungstates, in alloys, in fire-proofing fabrics and as a ceramic pigment. It is used to study their chromism, photocatalysis and sensing capabilities. It is associated with rhenium trioxide particles and used for detection of protonatable, radical and precharged species under laser desorption ionization.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | 1500°C to 1600°C (in N2) |
| Physical Form | Powder |
| Quantity | 5 g |
| Assay Percent Range | 99.9% (metals basis) |
| Solubility Information | Soluble in acid and potassium hydroxide. Insoluble in water. |
| Formula Weight | 215.85 |
| Density | 10.8 |
| Chemical Name or Material | Tungsten(IV) oxide |
Safety and Handling
| EINECSNumber | 234-842-7 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.