Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tungsten oxide, 99.99% (metals basis)

Catalog No. p-4904564
Click to view available options
Quantity:
50 g
250 g

CAS: 12037-58-0 | O2W | 215.84 g/mol

Tungsten oxide is used as a pigment in ceramics and paints. It is used in various types of infrared absorbing coatings and foils. It is involved in the production of electrochromic windows, which is an electrically switchable glass that change light transmission properties with an applied voltage. It is a unique material utilized for various applications due their chromism, photocatalysis, and sensing capabilities.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12037-58-0
Molecular Formula O2W
Molecular Weight (g/mol) 215.84
MDL Number MFCD00049709
InChI Key DZKDPOPGYFUOGI-UHFFFAOYSA-N
IUPAC Name dioxotungsten
SMILES O=[W]=O

Specifications

Melting Point 1500°C to 1600°C
Color Blue to Brown
Physical Form Powder
Quantity 50 g
Assay Percent Range 99.99% metal basis
Solubility Information Soluble in hot alkalis. Insoluble in water and acids.
Formula Weight 230.25
Density 7.16
Chemical Name or Material Tungsten oxide
Hazard Category H315-H319-H335
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA Yes

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.