Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tungsten, plasma standard solution, Specpure™ W 1000μg/mL

Catalog No. AA42248AE
Click to view available options
Quantity:
50 mL
100 mL
500 mL

Catalog No. AA42248AE

Supplier: Thermo Scientific Chemicals 042248AE

Only null left
Add to Cart

(NH4)2WO4 in H2O

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

Molecular Formula (NH4)2WO4 in H2O
MDL Number MFCD00011461

Specifications

Physical Form Liquid
Concentration W 1000 μg/mL
Quantity 100 mL
Chemical Name or Material Tungsten, plasma standard solution, Specpure™
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.