Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tungsten rod, 1.5mm (0.06in) dia, 99.95% (metals basis)

Catalog No. aa422339x
Click to view available options
Quantity:
30 cm
2 x 10 cm
5 x 30 cm

CAS: 7440-33-7 | W | 183.84 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7440-33-7
Molecular Formula W
Molecular Weight (g/mol) 183.84
MDL Number MFCD00011461
InChI Key WFKWXMTUELFFGS-UHFFFAOYSA-N
Synonym wolfram, ion, va, tungsten, elemental, wolframium, unii-v9306cxo6g, 4+ ion, tungsten, soluble compounds, tungsten, insoluble compounds, powder
PubChem CID 23964
ChEBI CHEBI:27998
IUPAC Name tungsten
SMILES [W]

Specifications

Quantity 2 x 10 cm
Weight ≈3.4g/10cm
Form Rod
Percent Purity 99.95%
Assay (metals basis)
Chemical Name or Material Tungsten rod, 1.5mm (0.06 in.) dia.
EINECSNumber 231-143-9
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.