Tungsten sulfide, 99.8% (metals basis), Alfa Aesar™

Manufacturer: Alfa Aesar 1182930

Quantity 250g

 View more versions of this product

Catalog No. AA1182930

  Check Availability
Add to Cart

    Chemical Identifiers

    Physical Form Powder
    Assay Percent Range 99.8% metal basis
    Molecular Formula WS2
    Formula Weight 247.98
    MDL Number MFCD00011467

    Specifications

    Formula Weight 247.98
    Quantity 250g
    Color Black
    Physical Form Powder
    Odor Odorless
    Assay Percent Range 99.8% metal basis
    Molecular Formula WS2
    MDL Number MFCD00011467

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit