Learn More
Tungsten(VI) oxide, Puratronic™, 99.998% (metals basis excluding Mo), Mo 100ppm
Description
Tungsten(VI) oxide is used in tungsten and tungstate manufacturing which are used as X-ray screens and for fire proofing fabrics. It is used as a ceramic pigment. Nanowires of tungsten(VI) oxide are capable of absorbing a higher percentage of the sun radiation since it absorbs blue light.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | 1473°C |
| Color | Yellow |
| Physical Form | Powder |
| Quantity | 25 g |
| Assay Percent Range | 99.998% metal basis excluding Mo |
| Solubility Information | Soluble in hot alkali solutions. Insoluble in water and acids. Slightly soluble in hydrofluoric acid. |
| Formula Weight | 231.85 |
| Odor | Odorless |
| Density | 7.16 |
| Chemical Name or Material | Tungsten(VI) oxide, Mo 100ppm |
Safety and Handling
| EINECSNumber | 215-231-4 |
| RTECSNumber | YO7760000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.