Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tunicamycin, 95%

Catalog No. aaj62217ma
Click to view available options
Quantity:
10 mg

CAS: 11089-65-9 | C39H64N4O16

Tunicamycin has been widely used in the study of glycoprotein synthesis in various biological systems. During protein glycosylation, tunicamycin is noted to be an inhibitor of the transfer of saccharide moieties to dolichol during dolichol-linked glycoprotein synthesis. Dose-dependent inhibition of DNA synthesis may be related to the alteration of glycoproteins, which thereby affects the transport of thymidine into cells. Additionally, tunicamycin has been reported to prevent cell cycle progression in primary cultures of rat glial cells, as well as inhibit lipid-mediated protein glycosylation in chick or mouse fibroblasts in a dose-dependent manner.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Tunicamycin has been widely used in the study of glycoprotein synthesis in various biological systems. During protein glycosylation, tunicamycin is noted to be an inhibitor of the transfer of saccharide moieties to dolichol during d
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 11089-65-9
Molecular Formula C39H64N4O16
MDL Number MFCD00065709

Specifications

Type Tunicamycin
Physical Form Powder
Color White to Yellow
Quantity 10 mg
Sensitivity Light and moisture sensitive
UN Number UN3462
Beilstein 6888090
Merck Index 14,9819
Solubility Information Soluble in methanol, ethanol, DMSO, or DMF
Formula Weight 844.95
Percent Purity 95%
Chemical Name or Material Tunicamycin
Show More Show Less
Hazard Category H300+H330-H373
Hazard Statement GHS H Statement
H300-H330-H373
Fatal if swallowed.
Fatal if inhaled.
May cause damage to organs through prolonged or repeated exposure.
Precautionary Statement P260-P264b-P270-P271-P284-P301+P310-P304+P340-P310-P314-P330-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: TOXINS, EXTRACTED FROM LIVING SOURCES, SOLID, N.O.S.
RTECSNumber YO7980200
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.