Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TUP1 Antibody (10), Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP266596
Description
TUP1 Monoclonal antibody specifically detects TUP1 in Yeast samples. It is validated for Western Blot.Specifications
TUP1 | |
Monoclonal | |
Unconjugated | |
PBS with 0.08% Sodium Azide | |
Alternative names, Flocculation suppressor protein, General transcriptional corepressor TUP1, Glucose repression regulatory protein TUP1, Repressor AER2 | |
Mouse | |
Protein A or G purified | |
Primary | |
Yeast | |
Purified |
Western Blot | |
10 | |
Western Blot 5-10 ug/ml | |
P16649 | |
TUP1 | |
Recombinant protein derived from the amino acids 1-200 of yeastt TUP1 protein (Unprot: P16649). | |
100 μg | |
850445 | |
Store at -20°C. Avoid freeze-thaw cycles. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction