Learn More
Invitrogen™ TUP1 Monoclonal Antibody (10)

Description
A suggested positive control for this product is Mav108 cells expressing recombinant TUP1 protein.
Specifications
Specifications
| Antigen | TUP1 |
| Applications | Western Blot |
| Classification | Monoclonal |
| Clone | 10 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.08% sodium azide |
| Gene | TUP1 |
| Gene Accession No. | P16649 |
| Gene Alias | AAR1; AER2; AMM1; chromatin-silencing transcriptional regulator TUP1; CRT4; CYC9; FLK1; Flocculation suppressor protein; General transcriptional corepressor TUP1; Glucose repression regulatory protein TUP1; Repressor AER2; ROX4; SFL2; TUP1; UMR7; YCR084C; YCR84C |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.