Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Turbidity Standard, 0 NTU (< 0.1 Nephelometric Turbidity Units), Ricca Chemical
Shop All Ricca Chemical Company Products

Click to view available options
Quantity:
500 mL
1 L
Specifications
Specifications
| Chemical Name or Material | Turbidity Standard, 0 NTU (<0.1 Nephelometric Turbidity Units) |
| Packaging | Amber Glass Bottle |
| CAS | 26628-22-8, 7732-18-5 |
| CAS Min % | 99.99, 0.01 |
| CAS Max % | 99.99, 0.01 |
| Density | 1 |
| Physical Form | Liquid |
| Color | Colorless |
| Melting Point | 0°C |
| Boiling Point | 100°C |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction