Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Turbidity Standard, 100 NTU, Polymer Bead Suspension, Ricca Chemical
SDP

Catalog No. r88060001c Shop All Ricca Chemical Company Products
Click to view available options
Quantity:
500 mL
1 L
4 L
Packaging:
Amber Glass Bottle
Clear PET-G Bottle

Chemical Identifiers

CAS 9003-53-6, 26628-22-8, 7732-18-5
Molecular Formula (C8H8)n
Molecular Weight (g/mol) 104.15
MDL Number MFCD00084450
InChI Key PPBRXRYQALVLMV-UHFFFAOYSA-N
PubChem CID 7501
ChEBI CHEBI:27452
IUPAC Name ethenylbenzene
SMILES *-CC(-*)C1=CC=CC=C1

Specifications

Packaging Amber Glass Bottle
CAS Min % 99.99, 0.01, 0.0047
CAS Max % 99.99, 0.01, 0.0047
Density 1
Physical Form Liquid
Color Colorless
Melting Point 0°C
Boiling Point 100°C
Solubility Miscible with water
Quantity 1 L
Chemical Name or Material Turbidity Standard, 100 NTU, Polymer Bead Suspension
Show More Show Less
ShelfLife 12 months
Recommended Storage Ambient
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.