Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Isopropylmagnesium Chloride Lithium Chloride Complex Solution

Catalog No. 656984800ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Click to view available options
Quantity:
100 mL
4 x 25 mL
800 mL
2 L
Danger

Catalog No. 65-698-4800ML

Supplier: Sigma Aldrich 656984800ML

Only null left
Add to Cart
Add to Cart
Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Reagent used with CH3MgCl/LiCl to affect magnesium-halogen exchange on unprotected aryl carboxylic acids.Sure/Seal is a trademark of Sigma-Aldrich Co. LLC

Specifications

Density 0.951 g/mL at 25°C
Molecular Formula C3H7Cl2LiMg
Linear Formula (CH3)2CHMgCl -+ LiCl
MDL Number MFCD07784514
Quantity 800 mL
Synonym Turbo Grignard
Molecular Weight (g/mol) 145.24 g/mol
Biological Activity Respiratory System
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.