Learn More
Thermo Scientific™ TurboFlow Cyclone Column
Catalog No. 03253870 Shop All Thermo Scientific Products
Encompass
Change view
Click to view available options
Material:
MAX
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Material
03-253-870 -
03-253-858 MAX
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 03-253-870 Supplier Thermo Scientific™ Supplier No. CH953282
Compatible with both Transcend and Aria TLX systems
TurboFlow Cyclone and TurboFlow XLcolumns work on both the Aria andTranscend TLX systems. They aremore robust and, for some applications, have a longer lifetime thanstandard TurboFlow columns.
TRUSTED_SUSTAINABILITY
Specifications
Diameter (Metric) 0.5 mm
Length (Metric) 50 mm
Stationary Phase C8