Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Honeywell Miller™ TurboLite™ Flash Personal Fall Limiters
Designed specifically for those who work at height and are exposed to arc flash hazards, welding, or who are working near high heat areas.
Supplier: Honeywell Miller™ MFLAF1/6FT
Description
- Engineered to deliver the assurance that is expected from Miller™
- TurboLite™ is an all-in-one solution for Arc flash and hot work applications
- Provides versatility, reliability, and maximum safety
- High impact housing with integral shock absorber
- Rated for up to a 420 lb. (189 kg) weight capacity for all connections down to foot level
- Meets the most stringent safety standards, including ANSI Z359.14 and ASTM F887
- Available in a wide range of lengths and configurations including single or twin versions, tie-back options, and products specifically designed for users working in bucket trucks

Specifications
Black, Yellow | |
Personal Fall Limiter | |
420 lb. | |
ANSI A10.32, ANSI Z359.14, ASTM F877, OSHA 1910.140, and OSHA 1926.502 | |
Arc Flash and Hot Work Applications | |
1 in. |
Single | |
Steel Locking Snap Hook, Steel Twist-Lock Carabiner | |
1 Ea. | |
Dupont™ Kevlar™™/Nomex™ | |
6 ft. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction