Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Honeywell Miller™ TurboLite™ Flash Personal Fall Limiters
Designed specifically for those who work at height and are exposed to arc flash hazards, welding, or who are working near high heat areas.
Specifications
Capacity (English) | 420 lb. |
---|---|
Certifications/Compliance | ANSI A10.32, ANSI Z359.14, ASTM F877, OSHA 1910.140, and OSHA 1926.502 |
Color | Black, Yellow |
Product Type | Personal Fall Limiter |
For Use With (Application) | Arc Flash and Hot Work Applications |
Catalog Number | Mfr. No. | Length (English) | Connector Types | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Length (English) | Connector Types | Price | Quantity & Availability | ||||
19-085-722
|
Honeywell Miller™
MFLAF212/6FT |
6 ft. | Quick-Connect Harness Connector and Aluminum Rebar Hook | N/A | N/A | N/A | |||
19-085-723
|
Honeywell Miller™
MFLAF23/6FT |
6 ft. | Quick-Connect Harness Connector and Steel Snap Hook | N/A | N/A | N/A | |||
19-085-724
|
Honeywell Miller™
MFLAFB1/6FT |
6 ft. | Locking Snap Hook and Aluminum Captive Eye Carabiner | N/A | N/A | N/A | |||
19-085-721
|
Honeywell Miller™
MFLAF1/6FT |
6 ft. | Steel Locking Snap Hook, Steel Twist-Lock Carabiner | N/A | N/A | N/A | |||
19-085-725
|
Honeywell Miller™
MFLAFT1/9FT |
9 ft. | Quick-Connect Harness Connector, Aluminum Captive Eye Carabiner, Steel Tie-Back Snap Hook (5 K) | N/A | N/A | N/A | |||
Description
- Engineered to deliver the assurance that is expected from Miller™
- TurboLite™ is an all-in-one solution for Arc flash and hot work applications
- Provides versatility, reliability, and maximum safety
- High impact housing with integral shock absorber
- Rated for up to a 420 lb. (189 kg) weight capacity for all connections down to foot level
- Meets the most stringent safety standards, including ANSI Z359.14 and ASTM F887
- Available in a wide range of lengths and configurations including single or twin versions, tie-back options, and products specifically designed for users working in bucket trucks
Specifications
420 lb. | |
Black, Yellow | |
Arc Flash and Hot Work Applications | |
Dupont™ Kevlar™™/Nomex™ |
ANSI A10.32, ANSI Z359.14, ASTM F877, OSHA 1910.140, and OSHA 1926.502 | |
Personal Fall Limiter | |
1 Ea. | |
1 in. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title