Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TurboLuc luciferase Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA1178
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA1178 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA1178 Supplier Invitrogen™ Supplier No. PA1178
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Western blot analysis of PA1-178 detects an ~16-18 kDa protein in Tluc expressing cells. PA1-178 can be used for activity-independent verification of Tluc expression in qIP assays.

TurboLuc™ (Tluc) is a novel intracellular luciferase derived from the marine copepod Metridia luciferase family. The Tluc gene was developed using directed mutagenesis for optimal performance and codon optimized for mammalian cell expression. Tluc is an ATP-independent enzyme that utilizes the substrate coelenterazine to produce intense bioluminescence light. Tluc is smaller and brighter than other commonly used luciferases, including firefly and Renilla. The intense bioluminescence greatly enhances sensitivity of Tluc luciferase-based assays, enabling detection of very minute amounts of luciferase (signal) in very sensitive assays such as protein interaction qIP assays (Prod # 82032, 82033, 82035, 82036).
TRUSTED_SUSTAINABILITY

Specifications

Antigen TurboLuc luciferase
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1mg/mL BSA, 30% glycerol and 0.05% sodium azide
Gene Alias TurboLuc luciferase synthetic
Host Species Rabbit
Immunogen Purified recombinant TurboLuc luciferase expressed in E. coli.
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Luciferase
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.