Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Turk Blood Diluting Fluid, for Leukocyte Count, Ricca Chemical
SDP

Catalog No. p-23609 Shop All Ricca Chemical Company Products
Click to view available options
Quantity:
500 mL
1 L
Packaging:
Natural Poly Bottle
This item is not returnable. View return policy

Catalog No. 8850-16

Supplier: Ricca Chemical Company 885016

Only null left
Add to Cart
Edge
Add to Cart
This item is not returnable. View return policy

For Analysis of Leucocyte Count

Specifications

Physical Form Solution
Chemical Name or Material Turk Blood Diluting Fluid
CAS 7732-18-5, 64-19-7, 548-62-9
CAS Min % 97.0, 1.01, 0.01
CAS Max % 100.0, 1.05, 0.01
Packaging Natural Poly Bottle
Color Dark Violet
Quantity 500 mL
Grade Laboratory
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.