Learn More
Description
Specifications
Specifications
| Antigen | IgG |
| Concentration | LYOPH |
| Conjugate | Biotin |
| Dilution | Restore with 1.0mL of deionized water (or equivalent). |
| Formulation | Lyophilized from 0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2, 10 mg/mL Bovine Serum Albumin (BSA) - Immunoglobulin and Protease free with 0.01% Sodium Azide |
| Gene Alias | Immunoglobulin G, ImmunoglobulinG |
| Gene Symbols | IGHG1 |
| Host Species | Turkey |
| Purification Method | Multi-step |
| Quantity | 1 mg |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
