Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TUSC1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA562408
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA562408 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA562408 Supplier Invitrogen™ Supplier No. PA562408
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: TNRRARDSGR EDEPGSPRAL RARLEKLEAM YRRALLQLHL EQRGPRPSGD KEEQPLQEPD SGLRSRDSEP S Highest antigen sequence identity to the following orthologs: Mouse - 70%, Rat - 33%.

This gene is located within the region of chromosome 9p that harbors tumor suppressor genes critical in carcinogenesis. It is an intronless gene which is downregulated in non-small-cell lung cancer and small-cell lung cancer cell lines, suggesting that it may play a role in lung tumorigenesis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TUSC1
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene TUSC1
Gene Accession No. Q2TAM9
Gene Alias 2200001D17Rik; TSG9; TSG-9; tumor suppressor candidate 1; tumor suppressor candidate gene 1 protein; Tumor suppressor candidate gene 1 protein homolog; tumour suppressor candidate 1; TUSC1
Gene Symbols TUSC1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human TUSC1. Recombinant protein control fragment (Product #RP-100015).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 286319
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.