Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TUSC1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA572128
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA572128 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA572128 Supplier Invitrogen™ Supplier No. PA572128
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

TUSC1 Polyclonal Antibody for Western Blot

This gene is located within the region of chromosome 9p that harbors tumor suppressor genes critical in carcinogenesis. It is an intronless gene which is downregulated in non-small-cell lung cancer and small-cell lung cancer cell lines, suggesting that it may play a role in lung tumorigenesis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TUSC1
Applications Western Blot
Classification Polyclonal
Concentration 0.45 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene TUSC1
Gene Accession No. Q673H1
Gene Alias 2200001D17Rik; TSG9; TSG-9; tumor suppressor candidate 1; tumor suppressor candidate gene 1 protein; Tumor suppressor candidate gene 1 protein homolog; tumour suppressor candidate 1; TUSC1
Gene Symbols TUSC1
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide between 111-138 amino acids from the Central region of mouse Tusc1.
Purification Method Antigen affinity chromatography, Protein A
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 69136
Target Species Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.