Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TUSC2 Monoclonal Antibody (1550CT388.74.12)
GREENER_CHOICE

Catalog No. PIMA524739
Change view
Click to view available options
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA524739 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA524739 Supplier Thermo Scientific Supplier No. MA524739
Add to Cart
Add to Cart

TUSC2 Monoclonal Antibody (1550CT388.74.12)

TUSC2 Monoclonal Antibody for Western Blot

TRUSTED_SUSTAINABILITY

Specifications

Antigen TUSC2
Applications Western Blot
Classification Monoclonal
Clone 1550CT388.74.12
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene TUSC2
Gene Accession No. O75896
Gene Alias 1190001E22Rik; AA407686; C3orf11; Fus1; Fus-1 protein; Fusion 1 protein; Lgcc; lung cancer candidiate, FUS; PAP; PDAP2; PDGFA associated protein 2; PDGFA-associated protein 2; Tumor suppressor candidate 2; TUSC 2; Tusc2
Gene Symbols TUSC2
Host Species Mouse
Immunogen human TUSC2 recombinant protein.
Purification Method Protein G
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11334
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.