Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TUSC2 Polyclonal Antibody

Rabbit Polyclonal Antibody

Supplier:  Invitrogen™ PA550882

Catalog No. PA550882


Only null left
Add to Cart

Description

Description

The antibody detects endogenous levels of total TUSC2 protein.

TUSC2 (tumor suppressor candidate 2), also known as FUS1, LGCC or PDAP2, is a 110 amino acid protein that is expressed at high levels in kidney, pancreas, lung, heart and skeletal muscle where it is thought to function as a tumor suppressor. More specifically, TUSC2 inhibits invasive colony formation by inducing G1 cell cycle arrest and ultimately causing apoptosis, thereby preventing tumor growth. The gene encoding TUSC2 maps to human chromosome 3, which houses over 1,100 genes, including a chemokine receptor (CKR) gene cluster and a variety of human cancer-related gene loci. Key tumor suppressing genes on chromosome 3 include those that encode the apoptosis mediator RASSF1, the cell migration regulator HYAL1 and the angiogenesis suppressor SEMA3B.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

TUSC2
Polyclonal
Unconjugated
TUSC2
1190001E22Rik; AA407686; C3orf11; Fus1; Fus-1 protein; Fusion 1 protein; Lgcc; lung cancer candidiate, FUS; PAP; PDAP2; PDGFA associated protein 2; PDGFA-associated protein 2; Tumor suppressor candidate 2; TUSC 2; Tusc2
Rabbit
Antigen affinity chromatography
RUO
11334
-20°C
Liquid
Immunohistochemistry (Paraffin)
2.9 mg/mL
PBS with 40% glycerol and 0.05% sodium azide; pH 7.4
O75896
TUSC2
Full length fusion protein of Human TUSC2.
100 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.