Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TUSC2 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501986736
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-6736 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-6736 Supplier Bioss Supplier No. BS5707R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

TUSC2 (tumor suppressor candidate 2), also known as FUS1, LGCC or PDAP2, is a 110 amino acid protein that is expressed at high levels in kidney, pancreas, lung, heart and skeletal muscle where it is thought to function as a tumor suppressor. More specifically, TUSC2 inhibits invasive colony formation by inducing G1 cell cycle arrest and ultimately causing apoptosis, thereby preventing tumor growth. The gene encoding TUSC2 maps to human chromosome 3, which houses over 1,100 genes, including a chemokine receptor (CKR) gene cluster and a variety of human cancer-related gene loci. Key tumor suppressing genes on chromosome 3 include those that encode the apoptosis mediator RASSF1, the cell migration regulator HYAL1 and the angiogenesis suppressor SEMA3B.

Specifications

Antigen TUSC2
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene TUSC2
Gene Accession No. O75896
Gene Alias 1190001E22Rik; AA407686; C3orf11; FUS1; Fus-1 protein; Fusion 1 protein; LGCC; lung cancer candidiate, FUS; PAP; Pdap2; PDGFA associated protein 2; PDGFA-associated protein 2; Tumor suppressor candidate 2; TUSC 2; TUSC2
Gene Symbols TUSC2
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human TUSC2.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11334
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.