Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ TUSC3 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA562109
Description
Immunogen sequence: GQKKKENLLA EKVEQLMEWS SRRSIFRMNG DKFRKFI Highest antigen sequence identity to the following orthologs: Mouse - 97%, Rat - 97%.
Made up of nearly 146 million bases, chromosome 8 encodes about 800 genes. Translocation of portions of chromosome 8 with amplifications of the c-Myc gene are found in some leukemias and lymphomas, and typically associated with a poor prognosis. Portions of chromosome 8 have been linked to schizophrenia and bipolar disorder. Trisomy 8, also known as Warkany syndrome 2, most often results in early miscarriage but is occasionally seen in a mosaic form in surviving patients who suffer to a varying degree from a number of symptoms including retarded mental and motor development, and certain facial and developmental defects. WRN is a DNA helicase encoded by chromosome 8 and shown defective in those with the early aging disorder Werner syndrome. Chromosome 8 is also associated with Pfeiffer syndrome, congenital hypothyroidism and Waardenburg syndrome.
Specifications
TUSC3 | |
Polyclonal | |
Unconjugated | |
TUSC3 | |
AU022242; BC003311; D8S1992; Dolichyl-diphosphooligosaccharide--protein glycosyltransferase subunit TUSC3; M33; Magnesium uptake/transporter TUSC3; MGC13453; MRT22; MRT7; N33; Oligosaccharyl transferase subunit TUSC3; oligosaccharyltransferase 3 homolog A; OST3A; Protein N33; Putative prostate cancer tumor suppressor; tumor suppressor candidate 3; Tusc3 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
7991 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q13454 | |
TUSC3 | |
Recombinant protein corresponding to Human TUSC3. Recombinant protein control fragment (Product #RP-103752). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction