Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TUSC3 Polyclonal Antibody
TUSC3 Polyclonal Antibody
Supplier: Thermo Scientific PA599274
Description
TUSC3 Polyclonal Antibody for Western Blot, ICC/IF

Specifications
TUSC3 | |
Polyclonal | |
Unconjugated | |
TUSC3 | |
AU022242; BC003311; D8S1992; Dolichyl-diphosphooligosaccharide--protein glycosyltransferase subunit TUSC3; M33; Magnesium uptake/transporter TUSC3; MGC13453; MRT22; MRT7; N33; Oligosaccharyl transferase subunit TUSC3; oligosaccharyltransferase 3 homolog A; OST3A; Protein N33; Putative prostate cancer tumor suppressor; tumor suppressor candidate 3; Tusc3 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
290783, 7991, 80286 | |
-20°C | |
Liquid |
Western Blot, Immunocytochemistry | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
Q13454, Q8BTV1 | |
TUSC3 | |
A synthesized peptide derived from human TUSC3(Accession Q13454), corresponding to amino acid residues Y133-A183. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction