Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TUT1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA541630
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA541630 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA541630 Supplier Invitrogen™ Supplier No. PA541630
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: EGAGGGAGTR AGWLATEAQV TQELKGLSGG EERPETEPLL SFVASVSPAD Sequence homology: Human: 100%.

TUT1 is a nucleotidyl transferase that functions as both a terminal uridylyltransferase and a nuclear poly(A) polymerase. TUT1 specifically adds and removes nucleotides from the 3' end of small nuclear RNAs and select mRNAs and may function in controlling gene expression and cell proliferation. TUT1 specifically catalyzes uridylylation of U6 snRNA (RNU6; MIM 180692) and is essential for cell proliferation (Trippe et al., 2006 [PubMed 16790842]).[supplied by OMIM].
TRUSTED_SUSTAINABILITY

Specifications

Antigen TUT1
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene TUT1
Gene Accession No. Q9H6E5
Gene Alias 2700038E08Rik; FLJ21850; FLJ22267; FLJ22347; MGC131987; MGC149809; nuclear speckle targeted phosphatidylinositol 4-phosphate 5-kinase type I-alpha regulated-poly(A) polymerase; nuclear speckle-targeted PIPK1A-regulated-poly(A) polymerase; PAP-associated domain-containing 2; PAPD2; poly(A) polymerase associated domain containing 2; RBM21; RNA binding motif protein 21; RNA uridylyltransferase; RNA-binding motif protein 21; RNA-binding protein 21; speckle targeted PIP5K1A-regulated poly(A) polymerase; STARPAP; Star-PAP; terminal uridylyl transferase 1 U6 snRNA-specific; terminal uridylyl transferase 1, U6 snRNA-specific; Tut1; TUTase; TUTase 6; TUTase6; U6 snRNA-specific terminal uridylyltransferase 1; U6 snRNA-specific terminal uridylyltransferase 1; speckle targeted PIP5K1A-regulated poly(A) polymerase; U6 TUTase; U6-TUTase; up-regulated in lung cancer 6; URLC6
Gene Symbols TUT1
Host Species Rabbit
Immunogen Synthetic peptide directed towards the middle region of human TUT1.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 64852
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.