Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TUT1 Rabbit anti-Rat, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP2860290.1ML
Description
TUT1 Polyclonal specifically detects TUT1 in Rat samples. It is validated for Western Blot.Specifications
TUT1 | |
Polyclonal | |
PBS, 2% Sucrose | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
64852 | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Western Blot | |
Unconjugated | |
EC 2.7.7.19, EC 2.7.7.52, FLJ21850, FLJ22267, FLJ22347, MGC131987, MGC149809, nuclear speckle targeted phosphatidylinositol 4-phosphate 5-kinase type I-alpharegulated-poly(A) polymerase, nuclear speckle-targeted PIPK1A-regulated-poly(A) polymerase, PAP-associated domain-containing 2, PAPD2, poly(A) polymerase associated domain containing 2, RBM21, RNA binding motif protein 21, RNA uridylyltransferase, RNA-binding motif protein 21, RNA-binding protein 21, speckle targeted PIP5K1A-regulated poly(A) polymerase, star-PAP, STARPAP, terminal uridylyl transferase 1, U6 snRNA-specific, TUTase, U6 snRNA-specific terminal uridylyltransferase 1, U6 TUTase, U6-TUTase | |
The immunogen is a synthetic peptide directed towards the N-terminal region of Rat TUT1 | |
0.1 mL | |
Primary | |
Rat | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction