Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TWA1 Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Supplier:  Invitrogen™ PA521785

Catalog No. PIPA521785


Only null left
Add to Cart

Description

Description

Recommended positive controls: 293T, A431, H1299, HeLaS3, HepG2, Molt-4, Raji. Predicted reactivity: Mouse (99%), Rat (99%), Xenopus laevis (96%), Chicken (97%), Rhesus Monkey (100%), Bovine (97%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

TWA1 was identified through a two hybrid-associated protein screen with RanBPM. TWA1 is well conserved through evolution and is localized within the nucleus. It interacts with RanBP9 and comprises a protein complex with RanBPM and Muskelin. TWA1 was found to possess the LisH-CTLH motif which is detected in proteins involved in microtubule dynamics, cell migration, nucleokinesis and chromosome segregation. These functions overlap with functions suggested for the RanGTPase cycle. Recent findings suggested that there is an as yet uncovered function of the RanGTPase cycle.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

TWA1
Polyclonal
Unconjugated
GID8
2310003C23Rik; 4833420G11Rik; AI451474; Bwk1; BWK-1; Bwk1 leukemia-related; C20orf11; GID complex subunit 8; GID complex subunit 8 homolog; GID complex subunit 8 homolog (S. cerevisiae); GID8; Glucose-induced degradation protein 8 homolog; protein C20orf11; protein C20orf11 homolog; TWA1; two hybrid associated protein 1; two hybrid associated protein No. 1 with RanBPM; two hybrid-associated protein 1 with RanBPM
Rabbit
Antigen affinity chromatography
RUO
296466, 54994
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
1 mg/mL
0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7
Q9NWU2
GID8
Recombinant protein encompassing a sequence within the center region of human C20orf11. The exact sequence is proprietary.
100 μL
Primary
Human, Rat
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

WARNING: Cancer - www.P65Warnings.ca.gov
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.