Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TWA1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA562096
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA562096 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA562096 Supplier Invitrogen™ Supplier No. PA562096
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: GQIQEAIALI NSLHPELLDT NRYLYFHLQQ QHLIELIRQR ETEAALEFAQ TQLAEQGEES RECLTEMERT LALLAF Highest antigen sequence identity to the following orthologs: Mouse - 100%, Rat - 100%.

TWA1 was identified through a two hybrid-associated protein screen with RanBPM. TWA1 is well conserved through evolution and is localized within the nucleus. It interacts with RanBP9 and comprises a protein complex with RanBPM and Muskelin. TWA1 was found to possess the LisH-CTLH motif which is detected in proteins involved in microtubule dynamics, cell migration, nucleokinesis and chromosome segregation. These functions overlap with functions suggested for the RanGTPase cycle. Recent findings suggested that there is an as yet uncovered function of the RanGTPase cycle.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TWA1
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene GID8
Gene Accession No. Q9NWU2
Gene Alias 2310003C23Rik; 4833420G11Rik; AI451474; Bwk1; BWK-1; Bwk1 leukemia-related; C20orf11; GID complex subunit 8; GID complex subunit 8 homolog; GID complex subunit 8 homolog (S. cerevisiae); GID8; Glucose-induced degradation protein 8 homolog; protein C20orf11; protein C20orf11 homolog; TWA1; two hybrid associated protein 1; two hybrid associated protein No. 1 with RanBPM; two hybrid-associated protein 1 with RanBPM
Gene Symbols GID8
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human GID8. Recombinant protein control fragment (Product #RP-99306).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54994
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.