Learn More
Invitrogen™ TWA1 Polyclonal Antibody

Description
A suggested positive control is human brain tissue lysate. PA5-20799 can be used with blocking peptide PEP-0913.
Specifications
Specifications
| Antigen | TWA1 |
| Applications | Immunohistochemistry, Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.02% sodium azide |
| Gene | GID8 |
| Gene Accession No. | Q9D7M1, Q9NWU2 |
| Gene Alias | 2310003C23Rik; 4833420G11Rik; AI451474; Bwk1; BWK-1; Bwk1 leukemia-related; C20orf11; GID complex subunit 8; GID complex subunit 8 homolog; GID complex subunit 8 homolog (S. cerevisiae); GID8; Glucose-induced degradation protein 8 homolog; protein C20orf11; protein C20orf11 homolog; TWA1; two hybrid associated protein 1; two hybrid associated protein No. 1 with RanBPM; two hybrid-associated protein 1 with RanBPM |
| Gene Symbols | GID8 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.