Learn More
Fisherbrand™ Twaddell Hydrometers
For use with liquids heavier than water such as milk, sulfuric acid and tanning solutions.
Description
- Permanently marked with individual serial numbers
Order Info
A certificate of calibration is available; simply add the letter C to the end of the catalog number when ordering.
Certifications
A certificate is provided to indicate traceability to standards provided by NIST™.
Compliance
Accuracy meets or exceeds ASTM™ E 100 specifications, Calibrated according to ASTM E 126 specifications.
Specifications
0.5° | |
290 mm | |
Liquids heavier than Water such as Milk, Sulfuric Acid, Tanning solutions |
3-point Cert, NIST, ASTM E100, E126 | |
300mL Borosilicate Glass cylinder | |
24 to 50° |